4-(Biphenyl-4-yl)pyridine

Supplier: Apollo Scientific
Warning

Synonyms: 4-(4-Biphenyl)pyridine

OR3583-1G OR3583-5G
APOSOR3583-1GEA 400 EUR
APOSOR3583-1G APOSOR3583-5G
4-(Biphenyl-4-yl)pyridine
4-(Biphenyl-4-yl)pyridine
Formula: C₁₇H₁₃N
MW: 231.292 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00657901
CAS Number: 861024-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR