3-Bromo-5-methyl-1H-pyrazolo[4,3-b]pyridine ≥98%
Supplier: Apollo Scientific
OR361743-100MG OR361743-1G
APOSOR361743-100MGEA 337 EUR
APOSOR361743-100MG APOSOR361743-1G
3-Bromo-5-methyl-1H-pyrazolo[4,3-b]pyridine ≥98%
3-Bromo-5-methyl-1H-pyrazolo[4,3-b]pyridine
Formula: C₇H₆BrN₃
MW: 212.047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257097
CAS Number: 52090-78-5