6-Iodobenzo[d][1,3]dioxol-5-amine ≥98%
Supplier: Apollo Scientific
OR370010-100MG OR370010-1G OR370010-250MG
APOSOR370010-100MGEA 91.5 EUR
APOSOR370010-100MG APOSOR370010-1G APOSOR370010-250MG
6-Iodobenzo[d][1,3]dioxol-5-amine ≥98%
6-Iodobenzo[d][1,3]dioxol-5-amine
Formula: C₇H₆INO₂
MW: 263.033 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23728743
CAS Number: 1000802-34-5