8-Ethynylisoquinoline ≥98%

Supplier: Apollo Scientific

OR370028-100MG OR370028-1G OR370028-500MG
APOSOR370028-100MGEA 598 EUR
APOSOR370028-100MG APOSOR370028-1G APOSOR370028-500MG
8-Ethynylisoquinoline ≥98%
8-Ethynylisoquinoline
Formula: C₁₁H₇N
Storage Temperature: Ambient
MDL Number: MFCD22665775
CAS Number: 1415559-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR