O-(2-Methylbenzyl)hydroxylamine hydrochloride ≥98%

Supplier: Apollo Scientific

OR370034-100MG
APOSOR370034-100MGEA 248 EUR
APOSOR370034-100MG
O-(2-Methylbenzyl)hydroxylamine hydrochloride ≥98%
O-(2-Methylbenzyl)hydroxylamine hydrochloride
Formula: C₈H₁₁NO·ClH
MW: 173.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18253006
CAS Number: 115777-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR