6-Bromobenzothiophen-3(2H)-one 1,1-dioxide ≥98%
Supplier: Apollo Scientific
OR370037-1G OR370037-250MG
APOSOR370037-1GEA 981 EUR
APOSOR370037-1G APOSOR370037-250MG
6-Bromobenzothiophen-3(2H)-one 1,1-dioxide ≥98%
6-Bromobenzothiophen-3(2H)-one 1,1-dioxide
Formula: C₈H₅BrO₃S
MW: 261.093 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27942487
CAS Number: 1240288-81-6