3-(3,4-Dichlorophenyl)-1H-pyrazole
Supplier: Apollo Scientific
Danger
OR38-1G OR38-250MG
APOSOR38-1GEA 139 EUR
APOSOR38-1G APOSOR38-250MG
3-(3,4-Dichlorophenyl)-1H-pyrazole
3-(3,4-Dichlorophenyl)-1H-pyrazole
Formula: C₉H₆Cl₂N₂
MW: 213.063 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01569519
CAS Number: 154257-68-8