Ethyl 1,4-dimethylpiperazine-2-carboxylate 97%
Supplier: Apollo Scientific
Danger
OR3900-1G OR3900-250MG
APOSOR3900-1GEA 52.2 EUR
APOSOR3900-1G APOSOR3900-250MG
Ethyl 1,4-dimethylpiperazine-2-carboxylate 97%
Ethyl 1,4-dimethylpiperazine-2-carboxylate
Formula: C₉H₁₈N₂O₂
MW: 186.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173923
CAS Number: 90729-01-4
UN: 1760
ADR: 8,III