5-Nitroisatoic anhydride 96%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Nitro-2H-3,1-benzoxazine-2,4(1H)-dione
OR400024-25G OR400024-100G
APOSOR400024-25GEA 150 EUR
APOSOR400024-25G APOSOR400024-100G
5-Nitroisatoic anhydride 96%
5-Nitroisatoic anhydride
Formula: C₈H₄N₂O₅
MW: 208.13 g/mol
Melting Pt: 265…266 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00016919
CAS Number: 4693-02-1