2-Bromo-6-iodonitrobenzene 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-3-iodo-2-nitrobenzene
OR400080-1G OR400080-250MG OR400080-5G
APOSOR400080-1GEA 63.8 EUR
APOSOR400080-1G APOSOR400080-250MG APOSOR400080-5G
2-Bromo-6-iodonitrobenzene 95%
2-Bromo-6-iodonitrobenzene
Formula: C₆H₃BrINO₂
MW: 327.902 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391758
CAS Number: 1126425-84-0