2-Bromo-3-methoxybenzyl alcohol
Supplier: Apollo Scientific
Warning
OR400088-1G OR400088-250MG
APOSOR400088-1GEA 308 EUR
APOSOR400088-1G APOSOR400088-250MG
2-Bromo-3-methoxybenzyl alcohol
2-Bromo-3-methoxybenzyl alcohol
Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391884
CAS Number: 199436-55-0