5,7-Dichlorobenzo[b]furan-3(2H)-one 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5,7-Dichloro-2,3-dihydro-3-oxobenzo[b]furan, 5,7-Dichloro-1-benzofuran-3(2H)-one
OR40012-1G OR40012-250MG
APOSOR40012-1GEA 406 EUR
APOSOR40012-1G APOSOR40012-250MG
5,7-Dichlorobenzo[b]furan-3(2H)-one 97%
5,7-Dichlorobenzo[b]furan-3(2H)-one
Formula: C₈H₄Cl₂O₂
MW: 203.022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08544411
CAS Number: 74815-20-6