2-Iodophenacyl bromide 95%
Supplier: Apollo Scientific
OR400132-1G OR400132-250MG OR400132-5G
APOSOR400132-1GEA 511 EUR
APOSOR400132-1G APOSOR400132-250MG APOSOR400132-5G
2-Iodophenacyl bromide 95%
2-Iodophenacyl bromide
Formula: C₈H₆BrIO
MW: 324.941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194409
CAS Number: 113258-84-7