4-Methoxy-3-nitrophenol
Supplier: Apollo Scientific
Warning
OR400153-1G OR400153-5G
APOSOR400153-1GEA 78.3 EUR
APOSOR400153-1G APOSOR400153-5G
4-Methoxy-3-nitrophenol
4-Methoxy-3-nitrophenol
Formula: C₇H₇NO₄
MW: 169.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00031284
CAS Number: 15174-02-4