3-(Methylsulphonyl)benzyl bromide 97%
Supplier: Apollo Scientific
Danger
OR40022-1G OR40022-250MG OR40022-5G
APOSOR40022-1GEA 0 EUR
APOSOR40022-1G APOSOR40022-250MG APOSOR40022-5G
3-(Methylsulphonyl)benzyl bromide 97%
Formula: C₈H₉BrO₂S
Melting Pt: 72…74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12025160
CAS Number: 82657-76-9
UN: 1759
ADR: 8,III