3-Methoxy-4-nitrobenzyl bromide
Supplier: Apollo Scientific
Danger
OR400222-250MG OR400222-1G
APOSOR400222-250MGEA 143 EUR
APOSOR400222-250MG APOSOR400222-1G
3-Methoxy-4-nitrobenzyl bromide
3-Methoxy-4-nitrobenzyl bromide
Formula: C₈H₈BrNO₃
MW: 246.058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08236862
CAS Number: 23145-65-5