3-Ethynylbenzoic acid 98%

Supplier: Apollo Scientific
Warning

OR400233-1G OR400233-5G
APOSOR400233-1GEA 24.8 EUR
APOSOR400233-1G APOSOR400233-5G
3-Ethynylbenzoic acid 98%
3-Ethynylbenzoic acid
Formula: C₉H₆O₂
Melting Pt: 164…171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06658448
CAS Number: 10601-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR