3-Bromo-5-chlorobenzyl alcohol
Supplier: Apollo Scientific
Synonyms: (3-Bromo-5-chlorophenyl)methanol
OR400319-25G OR400319-5G
APOSOR400319-25GEA 604 EUR
APOSOR400319-25G APOSOR400319-5G
3-Bromo-5-chlorobenzyl alcohol
3-Bromo-5-chlorobenzyl alcohol
Formula: C₇H₆BrClO
MW: 221.479 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061830
CAS Number: 917562-09-5