3'-Chloro-5'-methoxyacetophenone
Supplier: Apollo Scientific
Warning
OR400320-1G OR400320-250MG OR400320-5G
APOSOR400320-1GEA 406 EUR
APOSOR400320-1G APOSOR400320-250MG APOSOR400320-5G
3'-Chloro-5'-methoxyacetophenone
3'-Chloro-5'-methoxyacetophenone
Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22544395
CAS Number: 214760-34-6