Methyl 2-bromo-4-formylbenzoate
Supplier: Apollo Scientific
Warning
OR400422-250MG OR400422-1G
APOSOR400422-250MGEA 377 EUR
APOSOR400422-250MG APOSOR400422-1G
Methyl 2-bromo-4-formylbenzoate
Methyl 2-bromo-4-formylbenzoate
Formula: C₉H₇BrO₃
MW: 243.054 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26863464
CAS Number: 90484-52-9