Methyl 2-bromo-3-nitrobenzoate
Supplier: Apollo Scientific
Warning
OR400515-1G OR400515-25G OR400515-5G
APOSOR400515-1GEA 50.9 EUR
APOSOR400515-1G APOSOR400515-25G APOSOR400515-5G
Methyl 2-bromo-3-nitrobenzoate
Methyl 2-bromo-3-nitrobenzoate
Formula: C₈H₆BrNO₄
MW: 260.042 g/mol
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00087891
CAS Number: 5337-09-7