Methyl 2-bromo-3-iodobenzoate
Supplier: Apollo Scientific
Warning
OR400525-1G OR400525-5G OR400525-25G
APOSOR400525-1GEA 78.3 EUR
APOSOR400525-1G APOSOR400525-5G APOSOR400525-25G
Methyl 2-bromo-3-iodobenzoate
Methyl 2-bromo-3-iodobenzoate
Formula: C₈H₆BrIO₂
MW: 340.941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520418
CAS Number: 1261797-04-9