5-Bromo-2-iodo-4-methoxybenzoic acid 95%
Supplier: Apollo Scientific
Warning
OR400579-1G OR400579-250MG OR400579-5G
APOSOR400579-1GEA 364 EUR
APOSOR400579-1G APOSOR400579-250MG APOSOR400579-5G
5-Bromo-2-iodo-4-methoxybenzoic acid 95%
5-Bromo-2-iodo-4-methoxybenzoic acid
Formula: C₈H₆BrIO₃
MW: 356.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025501
CAS Number: 1269292-64-9