3'-Bromo-5'-methylacetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3-Bromo-5-methylphenyl)ethanone
OR400653-1G OR400653-5G
APOSOR400653-1GEA 143 EUR
APOSOR400653-1G APOSOR400653-5G
3'-Bromo-5'-methylacetophenone
3'-Bromo-5'-methylacetophenone
Formula: C₉H₉BrO
MW: 213.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18905445
CAS Number: 1379325-64-0