3-Methoxy-2-nitroaniline
Supplier: Apollo Scientific
Warning
OR400682-1G OR400682-5G OR400682-25G
APOSOR400682-1GEA 21.9 EUR
APOSOR400682-1G APOSOR400682-5G APOSOR400682-25G
3-Methoxy-2-nitroaniline
3-Methoxy-2-nitroaniline
Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD18324417
CAS Number: 16554-47-5