4-Methoxy-1H-benzimidazole 95+%
Supplier: Apollo Scientific
Synonyms: 4-Methoxy-1H-benzo[d]imidazole
OR400694-250MG OR400694-1G
APOSOR400694-250MGEA 27.7 EUR
APOSOR400694-250MG APOSOR400694-1G
4-Methoxy-1H-benzimidazole 95+%
4-Methoxy-1H-benzimidazole
Formula: C₈H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD08729266
CAS Number: 27080-53-1