Methyl 4-bromo-3-formylbenzoate
Supplier: Apollo Scientific
Warning
OR400768-1G OR400768-250MG
APOSOR400768-1GEA 101 EUR
APOSOR400768-1G APOSOR400768-250MG
Methyl 4-bromo-3-formylbenzoate
Methyl 4-bromo-3-formylbenzoate
Formula: C₉H₇BrO₃
MW: 243.054 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16036866
CAS Number: 858124-35-3