2-Bromo-6-methoxypyrazine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-6-methoxy-1,4-diazine
OR40079-1G OR40079-250MG OR40079-5G
APOSOR40079-1GEA 163 EUR
APOSOR40079-1G APOSOR40079-250MG APOSOR40079-5G
2-Bromo-6-methoxypyrazine 97%
2-Bromo-6-methoxypyrazine
Formula: C₅H₅BrN₂O
MW: 189.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10688549
CAS Number: 91678-76-1