(R)-4,5,6-Tetrahydro-3H-pyrrolo[1,2-c]oxathiazole 1,1-dioxide 95%
Supplier: Apollo Scientific
Warning
OR400805-1G OR400805-250MG OR400805-5G
APOSOR400805-1GEA 58 EUR
APOSOR400805-1G APOSOR400805-250MG APOSOR400805-5G
(R)-4,5,6-Tetrahydro-3H-pyrrolo[1,2-c]oxathiazole 1,1-dioxide 95%
(R)-4,5,6-Tetrahydro-3H-pyrrolo[1,2-c]oxathiazole 1,1-dioxide
Formula: C₅H₉NO₃S
MW: 163.195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26406787
CAS Number: 143577-46-2