2-Methoxy-3-methylbenzyl bromide
Supplier: Apollo Scientific
|
Warning
|
OR400877-250MG
OR400877-1G
APOSOR400877-250MGEA
267
EUR
APOSOR400877-250MG
APOSOR400877-1G
2-Methoxy-3-methylbenzyl bromide
2-Methoxy-3-methylbenzyl bromide
Formula:
C₉H₁₁BrO MW: 215.087 g/mol Boiling Pt: 131…140 °C (21 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD18398319 CAS Number: 122950-66-7 |