2-Bromo-3’-bromopropiophenone
Supplier: Apollo Scientific
Danger
OR400893-1G OR400893-250MG OR400893-5G
APOSOR400893-1GEA 212 EUR
APOSOR400893-1G APOSOR400893-250MG APOSOR400893-5G
2-Bromo-3’-bromopropiophenone
2-Bromo-3’-bromopropiophenone
Formula: C₉H₈Br₂O
MW: 291.967 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04118000
CAS Number: 76650-08-3