2-Bromo-6-chlorophenacyl bromide 95%
Supplier: Apollo Scientific
Danger
OR400928-1G OR400928-250MG
APOSOR400928-1GEA 671 EUR
APOSOR400928-1G APOSOR400928-250MG
2-Bromo-6-chlorophenacyl bromide 95%
2-Bromo-6-chlorophenacyl bromide
Formula: C₈H₅Br₂ClO
MW: 312.386 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18392521
CAS Number: 1261604-22-1
UN: 3261
ADR: 8 III