3,4-Dimethoxyphenylacetylene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,2-Dimethoxy-4-ethynylbenzene
OR40104-5G
APOSOR40104-5GEA 82.8 EUR
APOSOR40104-5G
3,4-Dimethoxyphenylacetylene 98%
3,4-Dimethoxyphenylacetylene
Formula: C₁₀H₁₀O₂
MW: 162.185 g/mol
Boiling Pt: 130 °C (15 mm Hg)
Melting Pt: 71…76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03839986
CAS Number: 4302-52-7