5-Bromo-2,3-dimethyl-2H-indazole
Supplier: Apollo Scientific
|
Warning
|
OR40138-100MG
OR40138-250MG
APOSOR40138-100MGEA
89.9
EUR
APOSOR40138-100MG
APOSOR40138-250MG
5-Bromo-2,3-dimethyl-2H-indazole
5-Bromo-2,3-dimethyl-2H-indazole
Formula:
C₉H₉BrN₂ MW: 225.085 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12028634 CAS Number: 878064-16-5 |