3-Chloro-6-methoxy-N-methylpyridine-2-carboxamide
Supplier: Apollo Scientific
|
Warning
|
OR40367-1G
OR40367-250MG
APOSOR40367-1GEA
145
EUR
APOSOR40367-1G
APOSOR40367-250MG
3-Chloro-6-methoxy-N-methylpyridine-2-carboxamide
3-Chloro-6-methoxy-N-methylpyridine-2-carboxamide
Formula:
C₈H₉ClN₂O₂ Melting Pt: 130…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD17676112 CAS Number: 1257535-58-2 |