(2S)-(+)-2-(Hydroxymethyl)tetrahydrofuran 96%
Supplier: Apollo Scientific
Warning
Synonyms: (S)-(+)-Tetrahydrofurfuryl alcohol, (S)-(Tetrahydrofuran-2-yl)methanol, (2S)-(+)-(Tetrahydrofur-2-yl)methanol
OR40670-1G OR40670-250MG OR40670-5G
APOSOR40670-1GEA 39.3 EUR
APOSOR40670-1G APOSOR40670-250MG APOSOR40670-5G
(2S)-(+)-2-(Hydroxymethyl)tetrahydrofuran 96%
(2S)-(+)-2-(Hydroxymethyl)tetrahydrofuran
Formula: C₅H₁₀O₂
MW: 102.132 g/mol
Boiling Pt: 80…83 °C
Storage Temperature: Ambient
MDL Number: MFCD04972320
CAS Number: 57203-01-7