2-Bromo-6-methylanisole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-2-methoxy-3-methylbenzene, 3-Bromo-2-methoxytoluene, 2-Bromo-6-methylphenyl methyl ether
OR40672-1G OR40672-5G OR40672-25G
APOSOR40672-1GEA 43.5 EUR
APOSOR40672-1G APOSOR40672-5G APOSOR40672-25G
2-Bromo-6-methylanisole 98%
2-Bromo-6-methylanisole
Formula: C₈H₉BrO
Boiling Pt: 62…63 °C (3 mm Hg)
Melting Pt: –39…–36 °C
Density: 1.41 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD12547891
CAS Number: 52200-69-8