2-Bromo-5-iodobenzaldehyde
Supplier: Apollo Scientific
Warning
OR40674-1G OR40674-5G OR40674-10G
APOSOR40674-1GEA 55.1 EUR
APOSOR40674-1G APOSOR40674-5G APOSOR40674-10G
2-Bromo-5-iodobenzaldehyde
2-Bromo-5-iodobenzaldehyde
Formula: C₇H₄BrIO
MW: 310.915 g/mol
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD07779027
CAS Number: 1032231-24-5