tert-Butyl (2R)-4-iodo-2-isopropyl-2,3-dihydropyrrolo[2,3-b]pyridine-1-carboxylate 95%
Supplier: Apollo Scientific
Warning
OR40693-1G OR40693-250MG
APOSOR40693-1GEA 92.8 EUR
APOSOR40693-1G APOSOR40693-250MG
tert-Butyl (2R)-4-iodo-2-isopropyl-2,3-dihydropyrrolo[2,3-b]pyridine-1-carboxylate 95%
tert-Butyl (2R)-4-iodo-2-isopropyl-2,3-dihydropyrrolo[2,3-b]pyridine-1-carboxylate
Formula: C₁₅H₂₁IN₂O₂
MW: 388.244 g/mol
Storage Temperature: Ambient
CAS Number: 2366996-94-1