tert-Butyl (2S)-4-iodo-2-isopropyl-2,3-dihydropyrrolo[2,3-b]pyridine-1-carboxylate 95%
Supplier: Apollo Scientific
Warning
OR40695-1G OR40695-250MG OR40695-5G
APOSOR40695-1GEA 111 EUR
APOSOR40695-1G APOSOR40695-250MG APOSOR40695-5G
tert-Butyl (2S)-4-iodo-2-isopropyl-2,3-dihydropyrrolo[2,3-b]pyridine-1-carboxylate 95%
tert-Butyl (2S)-4-iodo-2-isopropyl-2,3-dihydropyrrolo[2,3-b]pyridine-1-carboxylate
Formula: C₁₅H₂₁IN₂O₂
MW: 388.244 g/mol
Storage Temperature: Ambient
CAS Number: 2366997-14-8