9-(Bromomethyl)anthracene 95%

Supplier: Apollo Scientific
Danger

OR40727-5G OR40727-1G OR40727-250MG
APOSOR40727-5GEA 654 EUR
APOSOR40727-5G APOSOR40727-1G APOSOR40727-250MG
9-(Bromomethyl)anthracene 95%
9-(Bromomethyl)anthracene
Formula: C₁₅H₁₁Br
MW: 271.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00096212
CAS Number: 2417-77-8
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR