Methyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 99%
Supplier: Apollo Scientific
Warning
OR40775-1G OR40775-250MG
APOSOR40775-1GEA 101 EUR
APOSOR40775-1G APOSOR40775-250MG
Methyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 99%
Methyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate
Formula: C₁₄H₂₀BNO₄
MW: 277.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22494137
CAS Number: 844500-75-0