2-(1,3-Dioxolan-2-yl)aniline
Supplier: Apollo Scientific
Warning
OR40804-1G OR40804-250MG
APOSOR40804-1GEA 522 EUR
APOSOR40804-1G APOSOR40804-250MG
2-(1,3-Dioxolan-2-yl)aniline
2-(1,3-Dioxolan-2-yl)aniline
Formula: C₉H₁₁NO₂
MW: 165.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12828679
CAS Number: 26908-34-9