1-Bromo-4-prop-2-ynoxy-benzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-4-(prop-2-yn-1-yloxy)benzene
OR40819-10G OR40819-1G OR40819-5G
APOSOR40819-10GEA 696 EUR
APOSOR40819-10G APOSOR40819-1G APOSOR40819-5G
1-Bromo-4-prop-2-ynoxy-benzene 99%
1-Bromo-4-prop-2-ynoxy-benzene
Formula: C₉H₇BrO
MW: 211.055 g/mol
Boiling Pt: 144 °C (24 mm Hg)
Density: 1.468 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD09946935
CAS Number: 33133-45-8