4,5-Dimethylimidazole hydrochloride 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4,5-Dimethyl-1H-imidazole hydrochloride
OR41075-1G OR41075-5G
APOSOR41075-1GEA 109 EUR
APOSOR41075-1G APOSOR41075-5G
4,5-Dimethylimidazole hydrochloride 95%
4,5-Dimethylimidazole hydrochloride
Formula: C₅H₈N₂·ClH
MW: 132.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743540
CAS Number: 53316-51-1