N-Methyl-N-phenyl-2-(piperazin-1-yl)acetamide 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-{2-[Methyl(phenyl)amino]-2-oxoethyl}piperazine, {Methyl[(piperazin-1-yl)acetyl]amino]benzene
OR4185-10G OR4185-2G OR4185-5G
APOSOR4185-10GEA 168 EUR
APOSOR4185-10G APOSOR4185-2G APOSOR4185-5G
N-Methyl-N-phenyl-2-(piperazin-1-yl)acetamide 97%
N-Methyl-N-phenyl-2-(piperazin-1-yl)acetamide
Formula: C₁₃H₁₉N₃O
MW: 233.309 g/mol
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00040792
CAS Number: 318280-95-4