4-Amino-2-methoxy-5-nitropyridine
Supplier: Apollo Scientific
Warning
Synonyms: 2-Methoxy-5-nitropyridin-4-amine
OR42159-1G OR42159-250MG
APOSOR42159-1GEA 133 EUR
APOSOR42159-1G APOSOR42159-250MG
4-Amino-2-methoxy-5-nitropyridine
4-Amino-2-methoxy-5-nitropyridine
Formula: C₆H₇N₃O₃
MW: 169.138 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11977410
CAS Number: 127356-38-1