7-Bromo-4-chlorobenzofuro[3,2-d]pyrimidine
Supplier: Apollo Scientific
Warning
OR42275-1G OR42275-250MG
APOSOR42275-1GEA 654 EUR
APOSOR42275-1G APOSOR42275-250MG
7-Bromo-4-chlorobenzofuro[3,2-d]pyrimidine
7-Bromo-4-chlorobenzofuro[3,2-d]pyrimidine
Formula: C₁₀H₄BrClN₂O
MW: 283.509 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31714177
CAS Number: 2387695-88-5