4-Amino-3-cyclopropylpyridine
Supplier: Apollo Scientific
Warning
OR42288-1G OR42288-250MG OR42288-5G
APOSOR42288-1GEA 446 EUR
APOSOR42288-1G APOSOR42288-250MG APOSOR42288-5G
4-Amino-3-cyclopropylpyridine
4-Amino-3-cyclopropylpyridine
Formula: C₈H₁₀N₂
MW: 134.178 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20313017
CAS Number: 1338998-82-5