3-Iodobenzene-1,2-diamine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Iodophenylene-1,2-diamine, 1,2-Diamino-3-iodobenzene
OR43514-1G OR43514-250MG
APOSOR43514-1GEA 306 EUR
APOSOR43514-1G APOSOR43514-250MG
3-Iodobenzene-1,2-diamine 97%
3-Iodobenzene-1,2-diamine
Formula: C₆H₇IN₂
MW: 234.038 g/mol
Boiling Pt: 144 °C (5 mm Hg)
Melting Pt: 49…51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12028717
CAS Number: 34446-43-0